missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichloro-4-fluoro-1-nitrobenzene, 95%

Manufacturer:  Alfa Aesar™ H6132106

 View more versions of this product

Catalog No. AAH6132106



Specifications

Specifications

3,5-Dichloro-4-fluoro-1-nitrobenzene
5g
C6H2Cl2FNO2
1,3-dichloro-2-fluoro-5-nitro-benzene, 2,6-dichloro-4-nitrofluorobenzene, 3,5-dichloro-4-fluoro-1-nitrobenzene, 3,5-dichloro-4-fluoronitrobenzene, acmc-209hk6, benzene, 1,3-dichloro-2-fluoro-5-nitro, ksc495g6p, pubchem4387, zlchem 371
C1=C(C=C(C(=C1Cl)F)Cl)[N+](=O)[O-]
209.985
209.99
95%
White
3107-19-5
MFCD00129963
VMAATSFMXSMKPG-UHFFFAOYSA-N
1,3-dichloro-2-fluoro-5-nitrobenzene
76554
Powder

Chemical Identifiers

3107-19-5
209.985
VMAATSFMXSMKPG-UHFFFAOYSA-N
76554
C1=C(C=C(C(=C1Cl)F)Cl)[N+](=O)[O-]
C6H2Cl2FNO2
MFCD00129963
1,3-dichloro-2-fluoro-5-nitro-benzene, 2,6-dichloro-4-nitrofluorobenzene, 3,5-dichloro-4-fluoro-1-nitrobenzene, 3,5-dichloro-4-fluoronitrobenzene, acmc-209hk6, benzene, 1,3-dichloro-2-fluoro-5-nitro, ksc495g6p, pubchem4387, zlchem 371
1,3-dichloro-2-fluoro-5-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 221-466-3

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

Documents