missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichloro-4-fluoro-1-nitrobenzene, 95%

Manufacturer:  Alfa Aesar™ H6132122

 View more versions of this product

Catalog No. AAH6132122


 

Specifications

Specifications

3,5-Dichloro-4-fluoro-1-nitrobenzene
100g
95%
MFCD00129963
VMAATSFMXSMKPG-UHFFFAOYSA-N
1,3-dichloro-2-fluoro-5-nitrobenzene
76554
Powder
White
3107-19-5
C6H2Cl2FNO2
3,5-Dichloro-4-fluoronitrobenzene
C1=C(C=C(C(=C1Cl)F)Cl)[N+](=O)[O-]
209.985
209.99

Chemical Identifiers

3107-19-5
209.985
VMAATSFMXSMKPG-UHFFFAOYSA-N
76554
C6H2Cl2FNO2
MFCD00129963
3,5-Dichloro-4-fluoronitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C

SDS Documents