Please login to your online account to display your discounted pricing

3,5-Dichloro-4-(2-chloro-4-nitrophenoxy)benzene-1-sulfonyl chloride, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB03983EA

 View more versions of this product

Catalog No. BTB03983EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 417.05
Quantity 10g
Assay 97%
CAS Max % 100.0
Assay Percent Range 97%
CAS 175135-06-5
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Chemical Name or Material 3, 5-Dichloro-4-(2-chloro-4-nitrophenoxy)benzene-1-sulfonyl chloride
Packaging Amber Glass Bottle
Molecular Formula C12H5Cl4NO5S
MDL Number MFCD00052029
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3, 5-Dichloro-4-(2-chloro-4-nitrophenoxy)benzene-1-sulfonyl chloride
CAS 175135-06-5
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H5Cl4NO5S
Formula Weight 417.05
MDL Number MFCD00052029
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents