Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichloro-2-iodotoluene, 98%

$80.86 - $393.91

Chemical Identifiers

Chemical Name or Material 3,5-Dichloro-2-iodotoluene
CAS 175277-97-1
Molecular Formula C7H5Cl2I
Formula Weight 286.93g/mol
MDL Number MFCD00052856
Synonym 2,4-Dichloro-6-methyliodobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1145206

 
alfa aesar™
A11452-06
5g Each for $80.86

AAA1145214

 
alfa aesar™
A11452-14
25g Each for $393.91
Description & Specifications

Specifications

Melting Point 43° to 45°C
Quantity 5g
Chemical Name or Material 3,5-Dichloro-2-iodotoluene
CAS 175277-97-1
Assay 98%
Molecular Formula C7H5Cl2I
Formula Weight 286.93g/mol
MDL Number MFCD00052856
Synonym 2,4-Dichloro-6-methyliodobenzene

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.