Learn More
3,5-Dichloro-2-hydroxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
$453.29 - $453.29
Chemical Identifiers
CAS | 23378-88-3 |
---|---|
Molecular Formula | C6H3Cl3O3S |
Molecular Weight (g/mol) | 261.50 |
MDL Number | MFCD00007432 |
InChI Key | KXFQRJNVGBIDHA-UHFFFAOYSA-N |
Synonym | 3,5-dichloro-2-hydroxybenzenesulphonyl chloride, benzenesulfonyl chloride, 3,5-dichloro-2-hydroxy, 3,5-dichloro-2-hydroxybenzenesulfonylchloride, 3,5-dichloro-2-hydroxybenzene-1-sulfonyl chloride, 3,5-dichloro-2-hydroxy-benzenesulfonyl chloride, acmc-20andd, kxfqrjnvgbidha-uhfffaoysa, 2,4-dichloro-6-chlorosulfonyl phenol, 2,4-dichlorophenol-6-sulfonyl chloride, 3,5-dichloro-2-hydroxyphenyl chlorosulfone |
PubChem CID | 90078 |
IUPAC Name | 3,5-dichloro-2-hydroxybenzenesulfonyl chloride |
SMILES | OC1=C(Cl)C=C(Cl)C=C1S(Cl)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1214718
|
Thermo Scientific Chemicals
L1214718 |
50 g |
Each for $453.29
|
|
|||||
Description
3,5-Dichloro-2-hydroxybenzenesulfonyl chloride is used in the synthesis of (1R,2R)-(+)-1,2-(3,3?,5,5?-tetrachloro-2,2?-dihydroxydibenzenesulfonamido)-1,2-diphenylethane1 and (1R,2R)-(+)- 1,2-(3,3?,5,5?-tetrachloro-2,2?-dihydroxydibenzenesulfonamido)cyclohexane. It was used as chromogenic system in one-step kinetic method for the determination of 5?-nucleotidase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Dichloro-2-hydroxybenzenesulfonyl chloride is used in the synthesis of (1R,2R)-(+)-1,2-(3,3′,5,5′-tetrachloro-2,2′-dihydroxydibenzenesulfonamido)-1,2-diphenylethane1 and (1R,2R)-(+)- 1,2-(3,3′,5,5′-tetrachloro-2,2′-dihydroxydibenzenesulfonamido)cyclohexane. It was used as chromogenic system in one-step kinetic method for the determination of 5′-nucleotidase.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Incompatible with bases, moisture and oxidizing agents. Store under dry inert gas. Protect from humidity and water.
Chemical Identifiers
23378-88-3 | |
261.50 | |
KXFQRJNVGBIDHA-UHFFFAOYSA-N | |
90078 | |
OC1=C(Cl)C=C(Cl)C=C1S(Cl)(=O)=O |
C6H3Cl3O3S | |
MFCD00007432 | |
3,5-dichloro-2-hydroxybenzenesulphonyl chloride, benzenesulfonyl chloride, 3,5-dichloro-2-hydroxy, 3,5-dichloro-2-hydroxybenzenesulfonylchloride, 3,5-dichloro-2-hydroxybenzene-1-sulfonyl chloride, 3,5-dichloro-2-hydroxy-benzenesulfonyl chloride, acmc-20andd, kxfqrjnvgbidha-uhfffaoysa, 2,4-dichloro-6-chlorosulfonyl phenol, 2,4-dichlorophenol-6-sulfonyl chloride, 3,5-dichloro-2-hydroxyphenyl chlorosulfone | |
3,5-dichloro-2-hydroxybenzenesulfonyl chloride |
Specifications
23378-88-3 | |
C6H3Cl3O3S | |
50 g | |
2940993 | |
3,5-dichloro-2-hydroxybenzenesulphonyl chloride, benzenesulfonyl chloride, 3,5-dichloro-2-hydroxy, 3,5-dichloro-2-hydroxybenzenesulfonylchloride, 3,5-dichloro-2-hydroxybenzene-1-sulfonyl chloride, 3,5-dichloro-2-hydroxy-benzenesulfonyl chloride, acmc-20andd, kxfqrjnvgbidha-uhfffaoysa, 2,4-dichloro-6-chlorosulfonyl phenol, 2,4-dichlorophenol-6-sulfonyl chloride, 3,5-dichloro-2-hydroxyphenyl chlorosulfone | |
KXFQRJNVGBIDHA-UHFFFAOYSA-N | |
3,5-dichloro-2-hydroxybenzenesulfonyl chloride | |
90078 | |
97% |
78°C to 80°C | |
MFCD00007432 | |
UN3261 | |
Moisture sensitive | |
Hydrolyzes in water. | |
OC1=C(Cl)C=C(Cl)C=C1S(Cl)(=O)=O | |
261.50 | |
261.51 | |
3,5-Dichloro-2-hydroxybenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 245-619-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only