missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dichloro-2-hydroxybenzenesulfonyl chloride, 90-95%, ACROS Organics™

Manufacturer:  Acros Organics 163830250

 View more versions of this product

Catalog No. AC163830250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3, 5-Dichloro-2-hydroxybenzenesulfonyl chloride
Glass bottle
78°C to 84°C
90-95%
90.0
90-95%
Cl2C6H2(OH)SO2Cl
3,5-dichloro-2-hydroxybenzenesulphonyl chloride, benzenesulfonyl chloride, 3,5-dichloro-2-hydroxy, 3,5-dichloro-2-hydroxybenzenesulfonylchloride, 3,5-dichloro-2-hydroxybenzene-1-sulfonyl chloride, 3,5-dichloro-2-hydroxy-benzenesulfonyl chloride, acmc-20andd, kxfqrjnvgbidha-uhfffaoysa, 2,4-dichloro-6-chlorosulfonyl phenol, 2,4-dichlorophenol-6-sulfonyl chloride, 3,5-dichloro-2-hydroxyphenyl chlorosulfone
C1=C(C=C(C(=C1Cl)O)S(=O)(=O)Cl)Cl
261.497
261.51
≥90% (ex Cl)
Authentic
Beige to Brown
25g
23378-88-3
100.0
C6H3Cl3O3S
MFCD00007432
KXFQRJNVGBIDHA-UHFFFAOYSA-N
3,5-dichloro-2-hydroxybenzenesulfonyl chloride
90078
Powder

Chemical Identifiers

23378-88-3
261.497
KXFQRJNVGBIDHA-UHFFFAOYSA-N
90078
C1=C(C=C(C(=C1Cl)O)S(=O)(=O)Cl)Cl
C6H3Cl3O3S
MFCD00007432
3,5-dichloro-2-hydroxybenzenesulphonyl chloride, benzenesulfonyl chloride, 3,5-dichloro-2-hydroxy, 3,5-dichloro-2-hydroxybenzenesulfonylchloride, 3,5-dichloro-2-hydroxybenzene-1-sulfonyl chloride, 3,5-dichloro-2-hydroxy-benzenesulfonyl chloride, acmc-20andd, kxfqrjnvgbidha-uhfffaoysa, 2,4-dichloro-6-chlorosulfonyl phenol, 2,4-dichlorophenol-6-sulfonyl chloride, 3,5-dichloro-2-hydroxyphenyl chlorosulfone
3,5-dichloro-2-hydroxybenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 245-619-9

TSCA : TSCA

SDS Documents