Please login to your online account to display your discounted pricing

3,5-Dichloro-2-hydroxybenzenesulfonyl chloride, 90-95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 5-Dichloro-2-hydroxybenzenesulfonyl chloride
Name Note 90-95%
CAS 23378-88-3
CAS Min % 90.0
CAS Max % 100.0
Assay ≥90% (ex Cl)
Assay Percent Range 90-95%
Molecular Formula C6H3Cl3O3S
Linear Formula Cl2C6H2(OH)SO2Cl
Formula Weight 261.51
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC163830250

  SDS
thermo fisher scientific
163830250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥90% (ex Cl)
Physical Form Powder
Formula Weight 261.51
Quantity 25g
Color Light beige to light brown
EINECS Number 245-619-9
TSCA TSCA
Melting Point 78° to 84°C
CAS Min % 90.0
CAS Max % 100.0
Chemical Name or Material 3, 5-Dichloro-2-hydroxybenzenesulfonyl chloride
Packaging Glass bottle
Name Note 90-95%
Assay Percent Range 90-95%
Molecular Formula C6H3Cl3O3S
Linear Formula Cl2C6H2(OH)SO2Cl
MDL Number MFCD00007432
CAS 23378-88-3
Synonym 2, 4-Dichlorophenol-6-sulfonyl chloride
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.