Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichloro-2-fluoropyridine, 98%

$60.66 - $735.77

Chemical Identifiers

Chemical Name or Material 3,5-Dichloro-2-fluoropyridine
CAS 823-56-3
Molecular Formula C5H2Cl2FN
Formula Weight 165.98
MDL Number MFCD04039349
Beilstein 1365267
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2003903

 
alfa aesar™
L2003903
1g Each for $60.66

AAL2003906

 
alfa aesar™
L2003906
5g Each for $187.80

AAL2003914

 
alfa aesar™
L2003914
25g Each for $735.77
Description & Specifications

Specifications

Formula Weight 165.98
Melting Point 41° to 43°C
Chemical Name or Material 3,5-Dichloro-2-fluoropyridine
Assay 98%
MDL Number MFCD04039349
CAS 823-56-3
Beilstein 1365267
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Molecular Formula C5H2Cl2FN
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.