Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichloro-2',4'-dimethylbenzophenone, 97%

$61.57 - $227.81

Chemical Identifiers

Chemical Name or Material 3,5-Dichloro-2',4'-dimethylbenzophenone
CAS 306937-29-1
Molecular Formula C15H12Cl2O
Formula Weight 279.17
MDL Number MFCD00833417
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2185206

  SDS
alfa aesar™
B2185206
5g Each for $227.81

AAB2185203

  SDS
alfa aesar™
B2185203
1g Each for $61.57
Description & Specifications

Specifications

Formula Weight 279.17
Chemical Name or Material 3,5-Dichloro-2',4'-dimethylbenzophenone
Assay 97%
Molecular Formula C15H12Cl2O
MDL Number MFCD00833417
CAS 306937-29-1
Melting Point 55° to 58°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.