missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dichloro-2,4,6-trifluoropyridine, 97+%

$44.41 - $428.70

Chemical Identifiers

CAS 1737-93-5
Molecular Formula C5Cl2F3N
Molecular Weight (g/mol) 201.957
MDL Number MFCD00006226
InChI Key PKSORSNCSXBXOT-UHFFFAOYSA-N
Synonym 2,4,6-trifluoro-3,5-dichloropyridine, pyridine, 3,5-dichloro-2,4,6-trifluoro, 3,5-dichlorotrifluoropyridine, 2,6-dichloro-1,3,5-trifluorobenzene, 3,5-dichloro-2,4-6-trifluoropyridine, 2,4,6-trifluoro-3,5-dichloro pyridine, 3,5-dichloro-2,4,6-trifluoro-pyridine, ablock ab-12-8020, pubchem5003, acmc-1boj6
PubChem CID 61273
IUPAC Name 3,5-dichloro-2,4,6-trifluoropyridine
SMILES C1(=C(C(=NC(=C1Cl)F)F)Cl)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1020506 View Documents Alfa Aesar™
A1020506
5g Each for $44.41
 
AAA1020522 View Documents Alfa Aesar™
A1020522
100g Each for $428.70
 
AAA1020514 View Documents Alfa Aesar™
A1020514
25g Each for $147.27
 
Specifications

Chemical Identifiers

1737-93-5
201.957
PKSORSNCSXBXOT-UHFFFAOYSA-N
61273
C1(=C(C(=NC(=C1Cl)F)F)Cl)F
C5Cl2F3N
MFCD00006226
2,4,6-trifluoro-3,5-dichloropyridine, pyridine, 3,5-dichloro-2,4,6-trifluoro, 3,5-dichlorotrifluoropyridine, 2,6-dichloro-1,3,5-trifluorobenzene, 3,5-dichloro-2,4-6-trifluoropyridine, 2,4,6-trifluoro-3,5-dichloro pyridine, 3,5-dichloro-2,4,6-trifluoro-pyridine, ablock ab-12-8020, pubchem5003, acmc-1boj6
3,5-dichloro-2,4,6-trifluoropyridine

Specifications

159°C to 160°C
5g
C5Cl2F3N
UN2811
2,4,6-trifluoro-3,5-dichloropyridine, pyridine, 3,5-dichloro-2,4,6-trifluoro, 3,5-dichlorotrifluoropyridine, 2,6-dichloro-1,3,5-trifluorobenzene, 3,5-dichloro-2,4-6-trifluoropyridine, 2,4,6-trifluoro-3,5-dichloro pyridine, 3,5-dichloro-2,4,6-trifluoro-pyridine, ablock ab-12-8020, pubchem5003, acmc-1boj6
C1(=C(C(=NC(=C1Cl)F)F)Cl)F
201.957
201.96
3,5-Dichloro-2,4,6-trifluoropyridine
24°C to 26°C
1737-93-5
MFCD00006226
475874
PKSORSNCSXBXOT-UHFFFAOYSA-N
3,5-dichloro-2,4,6-trifluoropyridine
61273
≥97%
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H318-H315-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA