Please login to your online account to display your discounted pricing

3,5-Dichloro-1,2,4-thiadiazole, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3,5-Dichloro-1,2,4-thiadiazole
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C2Cl2N2S
Formula Weight 155.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC376030010

  SDS
thermo fisher scientific
AC376030010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC376030050

  SDS
thermo fisher scientific
AC376030050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC376030250

 
thermo fisher scientific
AC376030250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 3,5-Dichloro-1,2,4-thiadiazole
Color Clear colorless to slightly yellow
Quantity 1g
CAS 2254-88-8
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5580 to 1.5600 (20°C, 589nm)
Physical Form Liquid
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C2Cl2N2S
Formula Weight 155.01