missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Dichloro-1,2,4-thiadiazole, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 376030010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3,5-Dichloro-1,2,4-thiadiazole | |
165.0°C to 167.0°C | |
96% min. (GC) | |
C2Cl2N2S | |
1g | |
Solubility in water: slightly soluble in water | |
C1(=NSC(=N1)Cl)Cl | |
155.01 | |
155.01 |
2254-88-8 | |
Authentic | |
Glass bottle | |
1.5580 to 1.5600 | |
dichloro-1,2,4-thiadiazole, 3,5-dichloro-1,2,4 thiadiazole, 1,2,4-thiadiazole, 3,5-dichloro, 1,2,4-thiadiazole,3,5-dichloro, 3,5-dichloro-1,2,4 thiadiazol | |
CEUHPOVLEQUFCC-UHFFFAOYSA-N | |
3,5-dichloro-1,2,4-thiadiazole | |
2725055 | |
97% |
Chemical Identifiers
2254-88-8 | |
155.01 | |
dichloro-1,2,4-thiadiazole, 3,5-dichloro-1,2,4 thiadiazole, 1,2,4-thiadiazole, 3,5-dichloro, 1,2,4-thiadiazole,3,5-dichloro, 3,5-dichloro-1,2,4 thiadiazol | |
3,5-dichloro-1,2,4-thiadiazole |
C2Cl2N2S | |
CEUHPOVLEQUFCC-UHFFFAOYSA-N | |
2725055 | |
C1(=NSC(=N1)Cl)Cl |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only