Please login to your online account to display your discounted pricing

3,5-Dibromotoluene, 98%, ACROS Organics™

$66.84 - $66.84

Chemical Identifiers

Chemical Name or Material 3, 5-Dibromotoluene
Name Note 98%
CAS 1611-92-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7 H6 Br2
Linear Formula Br2C6H3CH3
Formula Weight 249.93
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC354350050

  SDS
thermo fisher scientific
35435-0050
Glass bottle 5g Each for $66.84
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 249.93
Physical Form Crystalline Solid
Quantity 5g
Color Slightly yellow
TSCA TSCA
Melting Point 37° to 41°C
CAS Max % 100.0
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Name Note 98%
Linear Formula Br2C6H3CH3
CAS 1611-92-3
MDL Number MFCD00013528
Assay Percent Range 98%
Molecular Formula C7 H6 Br2
Chemical Name or Material 3, 5-Dibromotoluene
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.