missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dibromothiophene-2-carboxylic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC446470010

 View more versions of this product

Catalog No. AC446470010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3, 5-Dibromothiophene-2-carboxylic acid
180°C to 188°C
7311-68-4
C5H2Br2O2S
YWOSIPRAZKDEIL-UHFFFAOYSA-N
3,5-dibromothiophene-2-carboxylic acid
625455
97%
Glass Bottle
1g
97%
2-thiophenecarboxylic acid, 3,5-dibromo, 3,5-dibromo-2-thiophenecarboxylic acid, 3,5-dibromo-2-thiophenecarboxylic acid #, 3,5-dibromo-thiophene-2-carboxylic acid, 3,5-dibromothiophene-2-carboxylic acid
C1=C(SC(=C1Br)C(=O)O)Br
285.94
285.94

Chemical Identifiers

7311-68-4
285.94
2-thiophenecarboxylic acid, 3,5-dibromo, 3,5-dibromo-2-thiophenecarboxylic acid, 3,5-dibromo-2-thiophenecarboxylic acid #, 3,5-dibromo-thiophene-2-carboxylic acid, 3,5-dibromothiophene-2-carboxylic acid
3,5-dibromothiophene-2-carboxylic acid
C5H2Br2O2S
YWOSIPRAZKDEIL-UHFFFAOYSA-N
625455
C1=C(SC(=C1Br)C(=O)O)Br
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents