missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Dibromothiophene-2-carboxylic acid, 97%, ACROS Organics™
Chemical Identifiers
CAS | 7311-68-4 |
---|---|
Molecular Formula | C5H2Br2O2S |
Molecular Weight (g/mol) | 285.94 |
InChI Key | YWOSIPRAZKDEIL-UHFFFAOYSA-N |
Synonym | 2-thiophenecarboxylic acid, 3,5-dibromo, 3,5-dibromo-2-thiophenecarboxylic acid, 3,5-dibromo-2-thiophenecarboxylic acid #, 3,5-dibromo-thiophene-2-carboxylic acid, 3,5-dibromothiophene-2-carboxylic acid |
PubChem CID | 625455 |
IUPAC Name | 3,5-dibromothiophene-2-carboxylic acid |
SMILES | C1=C(SC(=C1Br)C(=O)O)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC446470010
|
Thermo Fisher Scientific
AC446470010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC446470050
|
Thermo Fisher Scientific
AC446470050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
7311-68-4 | |
285.94 | |
2-thiophenecarboxylic acid, 3,5-dibromo, 3,5-dibromo-2-thiophenecarboxylic acid, 3,5-dibromo-2-thiophenecarboxylic acid #, 3,5-dibromo-thiophene-2-carboxylic acid, 3,5-dibromothiophene-2-carboxylic acid | |
3,5-dibromothiophene-2-carboxylic acid |
C5H2Br2O2S | |
YWOSIPRAZKDEIL-UHFFFAOYSA-N | |
625455 | |
C1=C(SC(=C1Br)C(=O)O)Br |
Specifications
Glass Bottle | |
1g | |
97% | |
2-thiophenecarboxylic acid, 3,5-dibromo, 3,5-dibromo-2-thiophenecarboxylic acid, 3,5-dibromo-2-thiophenecarboxylic acid #, 3,5-dibromo-thiophene-2-carboxylic acid, 3,5-dibromothiophene-2-carboxylic acid | |
C1=C(SC(=C1Br)C(=O)O)Br | |
285.94 | |
285.94 | |
3, 5-Dibromothiophene-2-carboxylic acid |
180°C to 188°C | |
7311-68-4 | |
C5H2Br2O2S | |
YWOSIPRAZKDEIL-UHFFFAOYSA-N | |
3,5-dibromothiophene-2-carboxylic acid | |
625455 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning