missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Dibromothiophene-2-carboxylic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC446470050
Specifications
3, 5-Dibromothiophene-2-carboxylic acid | |
5g | |
97% | |
2-thiophenecarboxylic acid, 3,5-dibromo, 3,5-dibromo-2-thiophenecarboxylic acid, 3,5-dibromo-2-thiophenecarboxylic acid #, 3,5-dibromo-thiophene-2-carboxylic acid, 3,5-dibromothiophene-2-carboxylic acid | |
C1=C(SC(=C1Br)C(=O)O)Br | |
285.94 | |
285.94 |
180°C to 188°C | |
7311-68-4 | |
C5H2Br2O2S | |
YWOSIPRAZKDEIL-UHFFFAOYSA-N | |
3,5-dibromothiophene-2-carboxylic acid | |
625455 | |
97% |
Chemical Identifiers
7311-68-4 | |
285.94 | |
2-thiophenecarboxylic acid, 3,5-dibromo, 3,5-dibromo-2-thiophenecarboxylic acid, 3,5-dibromo-2-thiophenecarboxylic acid #, 3,5-dibromo-thiophene-2-carboxylic acid, 3,5-dibromothiophene-2-carboxylic acid | |
3,5-dibromothiophene-2-carboxylic acid |
C5H2Br2O2S | |
YWOSIPRAZKDEIL-UHFFFAOYSA-N | |
625455 | |
C1=C(SC(=C1Br)C(=O)O)Br |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning