Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dibromosalicylic acid, 98%

$82.35 - $213.96

Chemical Identifiers

Chemical Name or Material 3,5-Dibromosalicylic acid
CAS 3147-55-5
Molecular Formula C7H4Br2O3
Formula Weight 295.92
MDL Number MFCD00002441
Beilstein 2210805
Merck Index 14,3028
Synonym 3,5-Dibromo-2-hydroxybenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2564214

 
alfa aesar™
B2564214
25g Each for $82.35

AAB2564222

 
alfa aesar™
B2564222
100g Each for $213.96
Description & Specifications

Specifications

Formula Weight 295.92
Chemical Name or Material 3,5-Dibromosalicylic acid
TSCA TSCA
Melting Point 223° to 228°C
Assay 98%
Quantity 25g
Molecular Formula C7H4Br2O3
MDL Number MFCD00002441
CAS 3147-55-5
Beilstein 2210805
Merck Index 14,3028
Synonym 3,5-Dibromo-2-hydroxybenzoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.