missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dibromosalicylaldehyde, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC153400010

 View more versions of this product

Catalog No. AC153400010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3, 5-Dibromosalicylaldehyde
80°C to 84°C
98%
Authentic
100.0
≥97.5%
C7H4Br2O2
MFCD00003318
JHZOXYGFQMROFJ-UHFFFAOYSA-N
3,5-dibromo-2-hydroxybenzaldehyde
7024
Crystalline Powder
Brown-Yellow to Yellow
1g
90-59-5
97.5
Glass bottle
98%
Br2C6H2(OH)CHO
3, 5-Dibromo-2-hydroxybenzaldehyde
C1=C(C=C(C(=C1Br)O)C=O)Br
279.915
279.91

Chemical Identifiers

90-59-5
279.915
JHZOXYGFQMROFJ-UHFFFAOYSA-N
7024
C7H4Br2O2
MFCD00003318
3, 5-Dibromo-2-hydroxybenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-003-4

RTECSNumber : CU5609000

TSCA : TSCA

SDS Documents