Please login to your online account to display your discounted pricing

3,5-Dibromopyridin-2-amine, 97%, Maybridge


Quantity 100g

Manufacturer: fisher scientific company  BTB07310FA

 View more versions of this product

Catalog No. BTB07310FA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 251.91
Quantity 100g
Assay 97%
CAS Max % 100.0
Linear Formula Br2(C5H2N)NH2
Molecular Formula C5H4Br2N2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3, 5-Dibromopyridin-2-amine
CAS Min % 96.5
MDL Number MFCD00038041
CAS 35486-42-1
Assay Percent Range 97%

Specifications

Chemical Name or Material 3, 5-Dibromopyridin-2-amine
CAS 35486-42-1
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H4Br2N2
Linear Formula Br2(C5H2N)NH2
Formula Weight 251.91
MDL Number MFCD00038041
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.