missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dibromobenzyl alcohol, 99%

$48.64 - $155.47

Chemical Identifiers

CAS 145691-59-4
Molecular Formula C7H6Br2O
Formula Weight 265.93
MDL Number MFCD01632143
Chemical Name or Material 3,5-Dibromobenzyl alcohol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2157403 SDS Alfa Aesar™
B2157403
1g Each for $48.64
AAB2157406 SDS Alfa Aesar™
B2157406
5g Each for $155.47
Specifications

Description

Applications
It is used in the synthesis of substituted benzylamines.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

145691-59-4
265.93
3,5-Dibromobenzyl alcohol
C7H6Br2O
MFCD01632143

Specifications

145691-59-4
1g
265.93
3,5-Dibromobenzyl alcohol
107° to 108°C
C7H6Br2O
MFCD01632143
99%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning