Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dibromobenzotrifluoride, 96%

Chemical Identifiers

Chemical Name or Material 3,5-Dibromobenzotrifluoride
CAS 401-84-3
Formula Weight 303.9
MDL Number MFCD00526219
Beilstein 2093912
Synonym 1,3-Dibromo-5-(trifluoromethyl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6142403

  SDS
alfa aesar™
H6142403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6142406

  SDS
alfa aesar™
H6142406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6142414

  SDS
alfa aesar™
H6142414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 303.9
Chemical Name or Material 3,5-Dibromobenzotrifluoride
Physical Form Liquid
Assay 96%
Color Pale yellow
Quantity 1g
MDL Number MFCD00526219
CAS 401-84-3
Beilstein 2093912
Synonym 1,3-Dibromo-5-(trifluoromethyl)benzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
3,5-Dibromobenzotrifluoride used as a intermediate in organic synthesis and pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 4°C. Storage: Keep container tightly closed. Store in a cool, dry and well-ventilated place.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.