missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dibromobenzotrifluoride, 96%

Manufacturer:  Alfa Aesar™ H6142414

 View more versions of this product

Catalog No. AAH6142414



Description

Description

Applications
3,5-Dibromobenzotrifluoride used as a intermediate in organic synthesis and pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 4°C. Storage: Keep container tightly closed. Store in a cool, dry and well-ventilated place.
Specifications

Specifications

3,5-Dibromobenzotrifluoride
25g
C7H3Br2F3
2093912
OFNXSUANJLHGQN-UHFFFAOYSA-N
1,3-dibromo-5-(trifluoromethyl)benzene
2063489
Liquid
Yellow
401-84-3
MFCD00526219
1,3-dibromo-5-trifluoromethyl ;benzene
C1=C(C=C(C=C1Br)Br)C(F)(F)F
303.904
303.9
96%

Chemical Identifiers

401-84-3
303.904
OFNXSUANJLHGQN-UHFFFAOYSA-N
2063489
C1=C(C=C(C=C1Br)Br)C(F)(F)F
C7H3Br2F3
MFCD00526219
1,3-dibromo-5-trifluoromethyl ;benzene
1,3-dibromo-5-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents