Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dibromobenzoic acid, 98%

$41.14 - $450.53

Chemical Identifiers

Chemical Name or Material 3,5-Dibromobenzoic acid
CAS 618-58-6
Molecular Formula C7H4Br2O2
Formula Weight 279.93
MDL Number MFCD00051758
Beilstein 1940691
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1665403

 
alfa aesar™
A1665403
1g Each for $41.14

AAA1665406

 
alfa aesar™
A1665406
5g Each for $116.32

AAA1665414

 
alfa aesar™
A1665414
25g Each for $450.53
Description & Specifications

Specifications

Formula Weight 279.93
Chemical Name or Material 3,5-Dibromobenzoic acid
Melting Point 216° to 224°C
Assay 98%
Quantity 1g
Molecular Formula C7H4Br2O2
MDL Number MFCD00051758
CAS 618-58-6
Beilstein 1940691
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DG6290010

Applications
3,5-Dibromobenzoic acid may be used for the syntheses (+)-menthyl 3,5-dibromobenzoate, di-tert-butyl 4-[2-(tert-butoxycarbonyl)ethyl]-4-(3,5-dibromobenzamido)heptanedioate2, (L)-methyl 2-(3,5-dibromobenzamido)-3-phenylpropanoate.

Solubility
Very faint turbidity in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.