Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dibromobenzamide, 97%

$61.11 - $823.51

Chemical Identifiers

Chemical Name or Material 3,5-Dibromobenzamide
CAS 175205-85-3
Molecular Formula C7H5Br2NO
Formula Weight 278.93
MDL Number MFCD00178771
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2494603

 
alfa aesar™
B2494603
1g Each for $61.11

AAB2494606

 
alfa aesar™
B2494606
5g Each for $204.73

AAB2494614

 
alfa aesar™
B2494614
25g Each for $823.51
Description & Specifications

Specifications

Formula Weight 278.93
Chemical Name or Material 3,5-Dibromobenzamide
Melting Point 180° to 184°C
Assay 97%
Quantity 1g
Molecular Formula C7H5Br2NO
MDL Number MFCD00178771
CAS 175205-85-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.