missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dibromobenzaldehyde +98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC393160010

 View more versions of this product

Catalog No. AC393160010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.0%
294.93
White to beige
100.0
C7H5Br2NO2
Authentic
609-85-8
Powder
1g
98.0
3, 5-Dibromoanthranilic acid
Glass bottle
98+%
MFCD00017087

Chemical Identifiers

609-85-8
100.0
C7H5Br2NO2
MFCD00017087
≥98.0%
98.0
98+%
294.93
3, 5-Dibromoanthranilic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.