Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dibromobenzaldehyde, 98%

$42.01 - $555.68

Chemical Identifiers

Chemical Name or Material 3,5-Dibromobenzaldehyde
CAS 56990-02-4
Molecular Formula C7H4Br2O
Formula Weight 263.93
MDL Number MFCD00156887
Beilstein 2573432
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2549503

 
alfa aesar™
B2549503
1g Each for $42.01

AAB2549506

 
alfa aesar™
B2549506
5g Each for $138.08

AAB2549514

 
alfa aesar™
B2549514
25g Each for $555.68
Description & Specifications

Specifications

Formula Weight 263.93
Chemical Name or Material 3,5-Dibromobenzaldehyde
Melting Point 89° to 93°C
Assay 98%
Quantity 1g
Molecular Formula C7H4Br2O
MDL Number MFCD00156887
CAS 56990-02-4
Beilstein 2573432
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.