missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dibromoanisole, 97%

Manufacturer:  Alfa Aesar™ H5592506

 View more versions of this product

Catalog No. AAH5592506



Specifications

Specifications

3,5-Dibromoanisole
34°C to 38°C
74137-36-3
MFCD02258848
OQZAQBGJENJMHT-UHFFFAOYSA-N
1,3-dibromo-5-methoxybenzene
11021812
97%
>110°C (230°F)
5g
C7H6Br2O
1,3-dibromo-5-methoxy-benzene
COC1=CC(=CC(=C1)Br)Br
265.932
265.93

Chemical Identifiers

74137-36-3
265.932
OQZAQBGJENJMHT-UHFFFAOYSA-N
11021812
COC1=CC(=CC(=C1)Br)Br
C7H6Br2O
MFCD02258848
1,3-dibromo-5-methoxy-benzene
1,3-dibromo-5-methoxybenzene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents