missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dibromo-o-phenylenediamine monohydrochloride, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 223735000

 View more versions of this product

Catalog No. AC223735000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
Powder
Beige
100.0
Authentic
3, 5-Dibromo-o-phenylenediamine monohydrochloride
99%
Br2C6H2(NH2)2·HCl
75568-11-5
302.4
500mg
13, 29
98.5
Glass bottle
99%
C6H6Br2N2·HCl
MFCD00012967

Chemical Identifiers

75568-11-5
100.0
C6H6Br2N2·HCl
302.4
3, 5-Dibromo-o-phenylenediamine monohydrochloride, 99%
98.5
99%
Br2C6H2(NH2)2·HCl
MFCD00012967
≥98.5%

Safety and Handling

SDS & Safety Documents

TSCA : TSCA