missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dibromo-N-(4-fluorobenzyl)aniline, 97%

Manufacturer:  Alfa Aesar™ H5747603

 View more versions of this product

Catalog No. AAH5747603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3,5-Dibromo-N-(4-fluorobenzyl)aniline
1858251-75-8
MFCD27975656
VBVKQLQBFBSBIU-UHFFFAOYSA-N
3,5-dibromo-N-[(4-fluorophenyl)methyl]aniline
97455621
Solid
1g
C13H10Br2FN
3,5-dibromo-n-4-fluorophenyl methyl aniline, 3,5-dibromo-n-4-fluorobenzyl aniline
C1=CC(=CC=C1CNC2=CC(=CC(=C2)Br)Br)F
359.036
359.03
97%

Chemical Identifiers

1858251-75-8
359.036
VBVKQLQBFBSBIU-UHFFFAOYSA-N
97455621
C1=CC(=CC=C1CNC2=CC(=CC(=C2)Br)Br)F
C13H10Br2FN
MFCD27975656
3,5-dibromo-n-4-fluorophenyl methyl aniline, 3,5-dibromo-n-4-fluorobenzyl aniline
3,5-dibromo-N-[(4-fluorophenyl)methyl]aniline
Documents