Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dibromo-N-(4-fluorobenzyl)aniline, 97%

Chemical Identifiers

Chemical Name or Material 3,5-Dibromo-N-(4-fluorobenzyl)aniline
Formula Weight 359.03
MDL Number MFCD27975656
Synonym N-(3,5-Dibromophenyl)-4-fluorobenzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH57476MD

 
alfa aesar™
H57476MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5747603

 
alfa aesar™
H5747603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 359.03
Chemical Name or Material 3,5-Dibromo-N-(4-fluorobenzyl)aniline
Physical Form Solid
Assay 97%
MDL Number MFCD27975656
Synonym N-(3,5-Dibromophenyl)-4-fluorobenzylamine
Quantity 250mg