missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dibromo-4-methylpyridine, 97%

$82.58 - $276.74

Chemical Identifiers

CAS 3430-23-7
Molecular Formula C6H5Br2N
Formula Weight 250.92
MDL Number MFCD00128859
Beilstein 1364818
Synonym 3,5-Dibromo-4-picoline
Chemical Name or Material 3,5-Dibromo-4-methylpyridine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL2003703 SDS Alfa Aesar™
L2003703
1g Each for $82.58
AAL2003706 SDS Alfa Aesar™
L2003706
5g Each for $276.74
Specifications Description & Specifications

Chemical Identifiers

3430-23-7
250.92
1364818
3,5-Dibromo-4-methylpyridine
C6H5Br2N
MFCD00128859
3,5-Dibromo-4-picoline

Specifications

3430-23-7
1g
250.92
1364818
3,5-Dibromo-4-methylpyridine
104° to 107°C
C6H5Br2N
MFCD00128859
3,5-Dibromo-4-picoline
97%

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger