Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dibromo-4-methylbenzoic acid, 98+%

$39.03 - $39.03

Chemical Identifiers

Chemical Name or Material 3,5-Dibromo-4-methylbenzoic acid
CAS 67973-32-4
Molecular Formula C8H6Br2O2
Formula Weight 293.94
MDL Number MFCD00017544
Beilstein 2577656
Synonym 3,5-Dibromo-p-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2539003

  SDS
alfa aesar™
B2539003
1g Each for $39.03

AAB2539006

  SDS
alfa aesar™
B2539006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 3,5-Dibromo-4-methylbenzoic acid
CAS 67973-32-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈242°C
Assay ≥98%
Quantity 1g
Molecular Formula C8H6Br2O2
Formula Weight 293.94
MDL Number MFCD00017544
Beilstein 2577656
Synonym 3,5-Dibromo-p-toluic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.