Please login to your online account to display your discounted pricing

3,5-Dibromo-4-hydroxybenzaldehyde, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  301170250

 View more versions of this product

Catalog No. AC301170250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 279.92
Physical Form Crystalline Powder or Needles
Quantity 25g
Color Pink to pale brown
TSCA TSCA
EINECS Number 221-017-1
Melting Point 182° to 186°C
CAS Max % 100.0
Packaging Glass bottle
CAS Min % 97.5
Infrared Spectrum Authentic
Chemical Name or Material 3, 5-Dibromo-4-hydroxybenzaldehyde
Assay Percent Range 98%
Molecular Formula C7H4Br2O2
Name Note 98%
MDL Number MFCD00016980
CAS 2973-77-5

Specifications

Chemical Name or Material 3, 5-Dibromo-4-hydroxybenzaldehyde
Name Note 98%
CAS 2973-77-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H4Br2O2
Formula Weight 279.92
MDL Number MFCD00016980