missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dibromo-4-hydroxybenzaldehyde, 98%, ACROS Organics™

Chemical Identifiers

CAS 2973-77-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H4Br2O2
Formula Weight 279.92
MDL Number MFCD00016980
Chemical Name or Material 3, 5-Dibromo-4-hydroxybenzaldehyde, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC301170250 SDS Thermo Fisher Scientific
301170250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC301170050 SDS Thermo Fisher Scientific
301170050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2973-77-5
100.0
C7H4Br2O2
MFCD00016980
≥97.5%
97.5
98%
279.92
3, 5-Dibromo-4-hydroxybenzaldehyde, 98%

Specifications

Pink to pale brown
25g
Authentic
100.0
Crystalline Powder or Needles
C7H4Br2O2
MFCD00016980
≥97.5%
182° to 186°C
2973-77-5
97.5
Glass bottle
98%
279.92
3, 5-Dibromo-4-hydroxybenzaldehyde, 98%

Safety and Handling

EINECSNumber : 221-017-1

TSCA : TSCA