missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dibromo-4-hydroxybenzaldehyde, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 301170050

 View more versions of this product

Catalog No. AC301170050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

3, 5-Dibromo-4-hydroxybenzaldehyde
182° to 186°C
98%
Authentic
100.0
Crystalline Powder or Needles
98%
279.92
Pink to pale brown
5g
2973-77-5
97.5
Glass bottle
≥97.5%
C7H4Br2O2
MFCD00016980

Chemical Identifiers

2973-77-5
100.0
C7H4Br2O2
MFCD00016980
≥97.5%
97.5
98%
279.92
3, 5-Dibromo-4-hydroxybenzaldehyde, 98%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 221-017-1

TSCA : TSCA