Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dibromo-4-hydroxybenzaldehyde, 98%

$49.11 - $218.49

Chemical Identifiers

Chemical Name or Material 3,5-Dibromo-4-hydroxybenzaldehyde
CAS 2973-77-5
Molecular Formula C7H4Br2O2
Formula Weight 279.93
MDL Number MFCD00016980
Beilstein 1948512
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1278006

  SDS
alfa aesar™
A1278006
5g Each for $49.11

AAA1278014

  SDS
alfa aesar™
A1278014
25g Each for $218.49
Description & Specifications

Specifications

Formula Weight 279.93
Chemical Name or Material 3,5-Dibromo-4-hydroxybenzaldehyde
Melting Point 179° to 182°C
Assay 98%
Quantity 5g
Molecular Formula C7H4Br2O2
MDL Number MFCD00016980
CAS 2973-77-5
Beilstein 1948512
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.