Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dibromo-2-hydroxypyridine, 97%

$53.49 - $687.08

Chemical Identifiers

Chemical Name or Material 3,5-Dibromo-2-hydroxypyridine
CAS 13472-81-6
Molecular Formula C5H3Br2NO
Formula Weight 252.89
MDL Number MFCD00023472
Beilstein 119389
Synonym 3,5-Dibromo-2-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1986203

  SDS
alfa aesar™
L1986203
1g Each for $53.49

AAL1986206

  SDS
alfa aesar™
L1986206
5g Each for $174.78

AAL1986214

  SDS
alfa aesar™
L1986214
25g Each for $687.08
Description & Specifications

Specifications

Formula Weight 252.89
Assay 97%
Molecular Formula C5H3Br2NO
MDL Number MFCD00023472
CAS 13472-81-6
Beilstein 119389
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 3,5-Dibromo-2-hydroxypyridine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈210°C
Quantity 1g
Odor Odorless
Synonym 3,5-Dibromo-2-pyridinol
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.