Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dibromo-2,6-dimethoxypyridine, 99%

$72.80 - $209.34

Chemical Identifiers

Chemical Name or Material 3,5-Dibromo-2,6-dimethoxypyridine
CAS 16727-44-9
Molecular Formula C7H7Br2NO2
Formula Weight 296.95
MDL Number MFCD00955954
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2759806

 
alfa aesar™
H2759806
5g Each for $72.80

AAH2759814

 
alfa aesar™
H2759814
25g Each for $209.34
Description & Specifications

Specifications

Formula Weight 296.95
Chemical Name or Material 3,5-Dibromo-2,6-dimethoxypyridine
Melting Point 85° to 87°C
Assay 99%
Quantity 5g
Molecular Formula C7H7Br2NO2
MDL Number MFCD00955954
CAS 16727-44-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.