Please login to your online account to display your discounted pricing

3,5-Dibromo-1H-1,2,4-triazole, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO07857CB

 View more versions of this product

Catalog No. MO07857CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 226.86
Quantity 250mg
Assay 97%
Chemical Name or Material 3,5-Dibromo-1H-1,2,4-triazole
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C2HBr2N3
CAS 7411-23-6

Specifications

Chemical Name or Material 3,5-Dibromo-1H-1,2,4-triazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C2HBr2N3
Formula Weight 226.86