Please login to your online account to display your discounted pricing

Alfa Aesar™ 3',5'-Dibenzyloxyacetophenone, 98%

$48.34 - $664.97

Chemical Identifiers

Chemical Name or Material 3',5'-Dibenzyloxyacetophenone
CAS 28924-21-2
Molecular Formula C22H20O3
Formula Weight 332.4
MDL Number MFCD00004777
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1961409

  SDS
alfa aesar™
A1961409
10g Each for $48.34

AAA1961418

  SDS
alfa aesar™
A1961418
50g Each for $160.08

AAA1961430

  SDS
alfa aesar™
A1961430
250g Each for $664.97
Description & Specifications

Specifications

Formula Weight 332.4
Chemical Name or Material 3',5'-Dibenzyloxyacetophenone
TSCA TSCA
Melting Point 60° to 62°C
Assay 98%
Quantity 10g
Molecular Formula C22H20O3
MDL Number MFCD00004777
CAS 28924-21-2

Applications
3,5-Dibenzyloxyacetophenone is used in the synthesis of enantiomerically pure B-ring modified analogs of (-)-epicatechin gallate.

Solubility
Soluble in dichloromethane, ethyl acetate and methanol

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.