Please login to your online account to display your discounted pricing

3,5-Diaminobenzoic Acid Dihydrochloride 99%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC151190100

 View more versions of this product

Catalog No. AC151190100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 225.08
Physical Form Fluffy Fine Crystalline Powder
Quantity 10g
Color Beige to gray
TSCA TSCA
RTECS Number DG6240000
EINECS Number 210-556-8
Beilstein 14, 453
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
CAS Min % 98.5
Packaging Glass bottle
Health Hazard 1 HYGROSCOPIC
Chemical Name or Material 3, 5-Diaminobenzoic acid dihydrochloride
Assay Percent Range 99%
Name Note 99%
Molecular Formula C7H8N2O2·2HCl
CAS 618-56-4
MDL Number MFCD00012983
Linear Formula (H2N)2C6H3CO22HCl

Specifications

Chemical Name or Material 3, 5-Diaminobenzoic acid dihydrochloride
Name Note 99%
CAS 618-56-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H8N2O2·2HCl
Linear Formula (H2N)2C6H3CO22HCl
Formula Weight 225.08
MDL Number MFCD00012983
Accessories

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents