Please login to your online account to display your discounted pricing

3,5-Diaminobenzoic Acid Dihydrochloride 99%, ACROS Organics™

$148.42 - $451.42

Chemical Identifiers

Chemical Name or Material 3, 5-Diaminobenzoic acid dihydrochloride
Name Note 99%
CAS 618-56-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H8N2O2·2HCl
Linear Formula (H2N)2C6H3CO22HCl
Formula Weight 225.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC151190100

  SDS
thermo fisher scientific
AC151190100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC151190250

  SDS
thermo fisher scientific
151190250
Glass bottle 25g Each for $148.42

AC151191000

  SDS
thermo fisher scientific
151191000
Glass bottle 100g Each for $451.42
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Fluffy Fine Crystalline Powder
Formula Weight 225.08
Quantity 10g
Color Beige to gray
EINECS Number 210-556-8
TSCA TSCA
RTECS Number DG6240000
Beilstein 14, 453
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material 3, 5-Diaminobenzoic acid dihydrochloride
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
CAS Min % 98.5
Health Hazard 1 HYGROSCOPIC
Molecular Formula C7H8N2O2·2HCl
Linear Formula (H2N)2C6H3CO22HCl
MDL Number MFCD00012983
CAS 618-56-4
Safety and Handling

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.