missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Diamino-1,2,4-triazole, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 112440050

 View more versions of this product

Catalog No. AC112440050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3,5-Diamino-1,2,4-triazole
Glass bottle
204°C to 208°C
1455-77-2
100.0
C2H5N5
26, 193
PKWIYNIDEDLDCJ-UHFFFAOYSA-N
1H-1,2,4-triazole-3,5-diamine
15078
99.09
98%
Authentic
White to Beige
5g
97.5
98%
MFCD00005233
1,2,4-triazolidine, 3,5-diimino, 3,5-diamino-1,2,4-triazole, 3,5-diamino-1h-1,2,4-triazole, 3,5-diamino-s-triazole, 3-amino-1h-1,2,4-triazol-5-ylamine, 4h-1,2,4-triazole-3,5-diamine, ccris 3745, guanazole, s-triazole, 3,5-diamino, unii-i01twm5267
C1(=NC(=NN1)N)N
99.09
CHEBI:75425
Crystalline Powder or Crystals

Chemical Identifiers

1455-77-2
99.09
PKWIYNIDEDLDCJ-UHFFFAOYSA-N
15078
1H-1,2,4-triazole-3,5-diamine
C2H5N5
MFCD00005233
1,2,4-triazolidine, 3,5-diimino, 3,5-diamino-1,2,4-triazole, 3,5-diamino-1h-1,2,4-triazole, 3,5-diamino-s-triazole, 3-amino-1h-1,2,4-triazol-5-ylamine, 4h-1,2,4-triazole-3,5-diamine, ccris 3745, guanazole, s-triazole, 3,5-diamino, unii-i01twm5267
CHEBI:75425
C1(=NC(=NN1)N)N
Safety and Handling

Safety and Handling

EINECSNumber : 215-937-2

SDS Documents