missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Diamino-1,2,4-triazole, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2277522

 View more versions of this product

Catalog No. AAB2277522



Description

Description

3,5-Diamino-1,2,4-triazole is used as an inhibitor of DNA synthesis. It also serves as an antitumor agents in the treatment of epigenetically-based diseases. It acts as a corrosion inhibitor for copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Diamino-1,2,4-triazole is used as an inhibitor of DNA synthesis. It also serves as an antitumor agents in the treatment of epigenetically-based diseases. It acts as a corrosion inhibitor for copper.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with acids and oxidizing agents.
Specifications

Specifications

3,5-Diamino-1,2,4-triazole
∼205°C (decomposition)
MFCD00005233
112467
Soluble in water.
C1(=NC(=NN1)N)N
99.097
CHEBI:75425
≥98%
1455-77-2
C2H5N5
100 g
guanazole, 3,5-diamino-1,2,4-triazole, 4h-1,2,4-triazole-3,5-diamine, 3,5-diamino-1h-1,2,4-triazole, 3,5-diamino-s-triazole, s-triazole, 3,5-diamino, 1,2,4-triazolidine, 3,5-diimino, unii-i01twm5267, ccris 3745, 3-amino-1h-1,2,4-triazol-5-ylamine
PKWIYNIDEDLDCJ-UHFFFAOYSA-N
1H-1,2,4-triazole-3,5-diamine
15078
99.1

Chemical Identifiers

1455-77-2
99.097
PKWIYNIDEDLDCJ-UHFFFAOYSA-N
15078
1H-1,2,4-triazole-3,5-diamine
C2H5N5
MFCD00005233
guanazole, 3,5-diamino-1,2,4-triazole, 4h-1,2,4-triazole-3,5-diamine, 3,5-diamino-1h-1,2,4-triazole, 3,5-diamino-s-triazole, s-triazole, 3,5-diamino, 1,2,4-triazolidine, 3,5-diimino, unii-i01twm5267, ccris 3745, 3-amino-1h-1,2,4-triazol-5-ylamine
CHEBI:75425
C1(=NC(=NN1)N)N
Safety and Handling

Safety and Handling

EINECSNumber : 215-937-2

RTECSNumber : XZ4535000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only