Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Diacetoxybenzoic acid, 97%

$78.04 - $261.68

Chemical Identifiers

Chemical Name or Material 3,5-Diacetoxybenzoic acid
CAS 35354-29-1
Molecular Formula C11H10O6
Formula Weight 238.2
MDL Number MFCD00017591
Beilstein 2741728
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1344103

  SDS
alfa aesar™
L1344103
1g Each for $78.04

AAL1344106

  SDS
alfa aesar™
L1344106
5g Each for $261.68
Description & Specifications

Specifications

Formula Weight 238.2
Chemical Name or Material 3,5-Diacetoxybenzoic acid
Molecular Formula C11H10O6
MDL Number MFCD00017591
CAS 35354-29-1
Beilstein 2741728
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 158° to 160°C
Quantity 1g
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.