missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Diacetoxybenzoic acid, 97%

Manufacturer:  Alfa Aesar™ L1344103

 View more versions of this product

Catalog No. AAL1344103



Specifications

Specifications

3,5-Diacetoxybenzoic acid
1g
C11H10O6
2741728
QBTDQJMLMVEUTQ-UHFFFAOYSA-N
3,5-diacetyloxybenzoic acid
539869
97%
158°C to 160°C
35354-29-1
MFCD00017591
3,5-diacetoxybenzoic acid, 3,5-bis acetyloxy benzoic acid, 3,5-diacethoxybenzoic acid, benzoic acid, 3,5-bis acetyloxy, 3,5-diacetoxy benzoic acid, maybridge1_004330, acmc-209igc, 3.5-diacetoxybenzoic acid, 3,5-diacetyloxybezoic acid, 3,5-diacetoxy-benzoic acid
CC(=O)OC1=CC(=CC(=C1)C(=O)O)OC(=O)C
238.195
238.2

Chemical Identifiers

35354-29-1
238.195
QBTDQJMLMVEUTQ-UHFFFAOYSA-N
539869
CC(=O)OC1=CC(=CC(=C1)C(=O)O)OC(=O)C
C11H10O6
MFCD00017591
3,5-diacetoxybenzoic acid, 3,5-bis acetyloxy benzoic acid, 3,5-diacethoxybenzoic acid, benzoic acid, 3,5-bis acetyloxy, 3,5-diacetoxy benzoic acid, maybridge1_004330, acmc-209igc, 3.5-diacetoxybenzoic acid, 3,5-diacetyloxybezoic acid, 3,5-diacetoxy-benzoic acid
3,5-diacetyloxybenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents