missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Di-tert-butylsalicylic acid, 97%

$36.31 - $118.78

Chemical Identifiers

CAS 19715-19-6
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.338
MDL Number MFCD00059058
InChI Key ZWQBZEFLFSFEOS-UHFFFAOYSA-N
Synonym 3,2-bis 1,1-dimethylethyl-2-hydroxybenzoic acid
PubChem CID 88208
IUPAC Name 3,5-ditert-butyl-2-hydroxybenzoic acid
SMILES CC(C)(C)C1=CC(=C(C(=C1)C(=O)O)O)C(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5510206 View Documents Alfa Aesar™
H5510206
5g Each for $36.31
 
AAH5510214 View Documents Alfa Aesar™
H5510214
25g Each for $118.78
 
Specifications

Chemical Identifiers

19715-19-6
250.338
ZWQBZEFLFSFEOS-UHFFFAOYSA-N
88208
CC(C)(C)C1=CC(=C(C(=C1)C(=O)O)O)C(C)(C)C
C15H22O3
MFCD00059058
3,2-bis 1,1-dimethylethyl-2-hydroxybenzoic acid
3,5-ditert-butyl-2-hydroxybenzoic acid

Specifications

179°C (354°F)
5g
C15H22O3
3,2-bis 1,1-dimethylethyl-2-hydroxybenzoic acid
CC(C)(C)C1=CC(=C(C(=C1)C(=O)O)O)C(C)(C)C
250.338
250.34
3,5-Di-tert-butylsalicylic acid
157°C to 162°C
19715-19-6
MFCD00059058
ZWQBZEFLFSFEOS-UHFFFAOYSA-N
3,5-ditert-butyl-2-hydroxybenzoic acid
88208
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 243-246-6

TSCA : Yes

Recommended Storage : Ambient temperatures