missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Di-tert-butylsalicylic acid, 97%

Manufacturer:  Alfa Aesar™ H5510214

 View more versions of this product

Catalog No. AAH5510214



Description

Description

Specifications

Specifications

3,5-Di-tert-butylsalicylic acid
157°C to 162°C
19715-19-6
MFCD00059058
ZWQBZEFLFSFEOS-UHFFFAOYSA-N
3,5-ditert-butyl-2-hydroxybenzoic acid
88208
97%
179°C (354°F)
25g
C15H22O3
3,2-bis 1,1-dimethylethyl-2-hydroxybenzoic acid, 3,5-bis-tert-butylsalicylic acid, 3,5-di-tert-butyl-2-hydroxybenzoic acid, 3,5-di-tert-butylsalicylic acid, 3,5-ditertbutyl salicylic acid, acmc-1c7qv, benzoic acid, 3,5-bis 1,1-dimethylethyl hydroxy, benzoic acid, 3,5-bis 1,1-dimethylethyl-2-hydroxy, dibutylsalicylicacid, ksc466o9n
CC(C)(C)C1=CC(=C(C(=C1)C(=O)O)O)C(C)(C)C
250.338
250.34

Chemical Identifiers

19715-19-6
250.338
ZWQBZEFLFSFEOS-UHFFFAOYSA-N
88208
CC(C)(C)C1=CC(=C(C(=C1)C(=O)O)O)C(C)(C)C
C15H22O3
MFCD00059058
3,2-bis 1,1-dimethylethyl-2-hydroxybenzoic acid, 3,5-bis-tert-butylsalicylic acid, 3,5-di-tert-butyl-2-hydroxybenzoic acid, 3,5-di-tert-butylsalicylic acid, 3,5-ditertbutyl salicylic acid, acmc-1c7qv, benzoic acid, 3,5-bis 1,1-dimethylethyl hydroxy, benzoic acid, 3,5-bis 1,1-dimethylethyl-2-hydroxy, dibutylsalicylicacid, ksc466o9n
3,5-ditert-butyl-2-hydroxybenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 243-246-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents