missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Di-tert-butylcatechol, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 112970050

 View more versions of this product

Catalog No. AC112970050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3, 5-Di-tert-butylcatechol
Glass bottle
White to Beige
5g
1020-31-1
100.0
C14H22O2
MFCD00008819
PJZLSMMERMMQBJ-UHFFFAOYSA-N
3,5-ditert-butylbenzene-1,2-diol
66099
Needle-Like Crystalline Powder
Technical
Authentic
>1000°C
97°C to 100°C
99%
98.5
99%
[(CH3)3C]2C6H2(OH)2
1,2-benzenediol, 3,5-bis 1,1-dimethylethyl, 1,2-benzenediol, 3,5-di 1,1-dimethylethyl, 3,5-bis tert-butyl benzene-1,2-diol, 3,5-di-t-butylcatechol, 3,5-di-tert-butylbenzene-1,2-diol, 3,5-di-tert-butylcatechol, 3,5-di-tert-butylpyrocatechol, 4,6-di-tert-butylpyrocatechol, pyrocatechol, 3,5-di-tert-butyl, unii-1328kn3f8b
CC(C)(C)C1=CC(=C(C(=C1)O)O)C(C)(C)C
222.33
222.33
99%

Chemical Identifiers

1020-31-1
222.33
PJZLSMMERMMQBJ-UHFFFAOYSA-N
66099
CC(C)(C)C1=CC(=C(C(=C1)O)O)C(C)(C)C
C14H22O2
MFCD00008819
1,2-benzenediol, 3,5-bis 1,1-dimethylethyl, 1,2-benzenediol, 3,5-di 1,1-dimethylethyl, 3,5-bis tert-butyl benzene-1,2-diol, 3,5-di-t-butylcatechol, 3,5-di-tert-butylbenzene-1,2-diol, 3,5-di-tert-butylcatechol, 3,5-di-tert-butylpyrocatechol, 4,6-di-tert-butylpyrocatechol, pyrocatechol, 3,5-di-tert-butyl, unii-1328kn3f8b
3,5-ditert-butylbenzene-1,2-diol
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 213-816-9

TSCA : TSCA

SDS Documents