missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Di-tert-butylbenzoic acid, 99%

Manufacturer:  Alfa Aesar™ L1238603

 View more versions of this product

Catalog No. AAL1238603



Description

Description

Applications
3,5-Di-tert-butylbenzoic acid is used in the synthesis of 5,5-di-tert-butylanilinium sulfate.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
Specifications

Specifications

3,5-Di-tert-butylbenzoic acid
1g
C15H22O2
981248
NCTSLPBQVXUAHR-UHFFFAOYSA-N
3,5-ditert-butylbenzoic acid
85339
99%
160°C to 162°C
16225-26-6
MFCD00082727
3,5-di-tert-butylbenzoic acid, 3,5-di-t-butylbenzoic acid, 3,5-ditertbutylbenzoic acid, 3,5-di-tert-butylbenzoicacid, 3,5-bis tert-butyl benzoic acid, benzoic acid, 3,5-bis 1,1-dimethylethyl, pubchem4953, acmc-209do2, 3,5-di-tertbutylbenzoic acid, ksc494i2n
CC(C)(C)C1=CC(=CC(=C1)C(=O)O)C(C)(C)C
234.339
234.34

Chemical Identifiers

16225-26-6
234.339
NCTSLPBQVXUAHR-UHFFFAOYSA-N
85339
CC(C)(C)C1=CC(=CC(=C1)C(=O)O)C(C)(C)C
C15H22O2
MFCD00082727
3,5-di-tert-butylbenzoic acid, 3,5-di-t-butylbenzoic acid, 3,5-ditertbutylbenzoic acid, 3,5-di-tert-butylbenzoicacid, 3,5-bis tert-butyl benzoic acid, benzoic acid, 3,5-bis 1,1-dimethylethyl, pubchem4953, acmc-209do2, 3,5-di-tertbutylbenzoic acid, ksc494i2n
3,5-ditert-butylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents